EMAIL THIS PAGE TO A FRIEND
Life sciences
Role of licochalcone C in cardioprotection against ischemia/reperfusion injury of isolated rat heart via antioxidant, anti-inflammatory, and anti-apoptotic activities.
PMID 25921769
Abstract
This study aimed to evaluate the protective effect of licochalcone C against myocardial ischemia/reperfusion injury in rats. Left ventricular developed pressure (LVDP) and its maximum up/down rate (±dp/dtmax) were recorded as myocardial function. Levels of creatine kinase (CK), lactate dehydrogenase (LDH), malondialdehyde (MDA), superoxide dismutase (SOD), glutathione/glutathione disulfide (GSH/GSSG) ratio, and tumor necrosis factor-alpha (TNF-α) were determined by using enzyme-linked immunosorbent assay. Cell morphology was observed and mitochondrial damage was assessed by HE coloration and transmission electron microscopy, respectively. Cardiomyocyte apoptosis was determined by using terminal deoxynucleotidyl transferased UTP nick-end labeling (TUNEL). Pretreatment with licochalcone C significantly improved the recovery of LVDP and ±dp/dtmax, and increased the levels of SOD and GSH/GSSG ratio. However, pretreatment with licochalcone C not only decreased the TUNEL-positive cell ratio and morphological changes, but also weaken the mitochondrial injury and the levels of CK, LDH, MDA, and TNF-α. These results suggested an important function of licochalcone C extracted from traditional Chinese medicine in the cardioprotection via antioxidant, anti-inflammatory, and anti-apoptotic activities.