PloS one
Metabolic and pancreatic effects of bone marrow mesenchymal stem cells transplantation in mice fed high-fat diet.
PMID 25923733
Abstract
The purpose of this study was to investigate the effects of multiple infusions of allogeneic MSCs on glucose homeostasis and morphometry of pancreatic islets in high- fat diet (HFD) fed mice. Swiss mice were fed standard diet (C group) or HFD (HFD group). After 8 weeks, animals of HFD group received sterile phosphate-buffered saline infusions (HFD-PBS) or four infusions of MSCs one week apart (HFD-MSCs). Fasting glycemia (FG) was determined weekly and glucose (GTT) and insulin (ITT) tolerance tests were performed 4, 8, 12, and 16 weeks after the infusions of MSCs. The MSCs transplanted mice were classified as responder (FG < 180 mg/dL, 72.2% of transplanted mice) or non-responder (FG > 180mg/dL, 28.8%) Seven weeks after MSCs infusions, FG decreased in HFD-MSCs responder mice compared with the HFD-PBS group. Sixteen weeks post MSCs infusions, GTT and ITT areas under the curve (AUC) decreased in HFD-MSCs responder mice compared to HFD-PBS group. Serum insulin concentration was higher in HFD-PBS group than in control animals and was not different compared with the other groups. The relative volume of α-cells was significantly smaller in HFD-PBS group than in C group and significantly higher in HFD-MSCs-NR than in HFD-PBS and HFD-MSCs-R groups. Cell apoptosis in the islets was higher in HFD-PBS group than in C group, and lower in HFD-MSCs responder mice than in HFD-PBS group and non-responder animals. The results demonstrate the ability of multiple infusions of MSCs to promote prolonged decrease in hyperglycemia and apoptosis in pancreatic islets and increase in insulin sensitivity in HFD fed mice.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2044
Glucagon, synthetic, powder, cell culture tested
C153H225N43O49S
G1774
Glucagon, ≥95% (HPLC), powder, synthetic
C153H225N43O49S