EMAIL THIS PAGE TO A FRIEND
International journal of surgery (London, England)
The effects of sulforaphane on the liver and remote organ damage in hepatic ischemia-reperfusion model formed with pringle maneuver in rats.
PMID 25924817
Abstract
The purpose of this study was to investigate the effect of Sulforaphane on ischemia/ reperfusion (IR) injury of the liver and distant organs resulting from liver blood flow arrest. Fourty Wistar rats were assigned into four groups, each included 10 rats were used. Group I as only laparatomy, Group II laparatomy and Sulforaphane application, Group III hepatic IR; and Group IV as hepatic IR and Sulforaphane application group. Animals were subjected to liver ischemia for 30 min and then reperfusion is started. 5 mg/kg Sulforaphane was applied via oral lavage 15 minutes before initiating the experimental study. Blood samples were taken from the animals for biochemical analysis at 60th minutes of the experiment in the first and second groups; 30 minutes after beginning reperfusion in the third and forth groups. Simultaneously, liver, lung and kidney tissues were sampled for biochemical and histopathological examinations. The administration of sulforaphane significantly reduced the serum TOA and liver TOA levels, increased the serum TAC and liver TAC levels and also decreased The OSI and liver OSI levels. In the histopathologic examination, the injury was reduced by the administration of sulforaphane. Administration of sulforaphane did not lead to any significant changes in any parameter including histopathological parameters in both the kidney and the lung. Sulforaphane reduced the liver oxidative stress from I/R injury. A histological injury in liver was reduced by sulforaphane administration. However, there were no significant effects of sulforaphane on the remote organ injuries induced by IR.