EMAIL THIS PAGE TO A FRIEND
International journal of experimental pathology
Influence of dietary supplementation with flaxseed and lactobacilli on the mucosal morphology and proliferative cell rate in the jejunal mucosa of piglets after weaning.
PMID 25929724
Abstract
The aim of the study was to investigate the influence of flaxseed and lactobacilli supplementation to the diet of piglets during the time period between 10xa0days before and 21xa0days after weaning. The morphometry of the jejunal mucosa and proliferative ratio of both epithelial and lamina propria cells were compared with those found in a group of piglets fed with the usual diet added with sunflower oil during the same time period. The addition of flaxseed oil to the diet significantly increased the crypt depth in comparison with both groups supplemented with sunflower (Pxa0