EMAIL THIS PAGE TO A FRIEND
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft
Monetite granules versus particulate autologous bone in bone regeneration.
PMID 25929816
Abstract
The aim of this study was to test bone tissue response to monetite granules in comparison with intramembranous autologous bone graft in a rabbit calvaria critical size defect model. Novel monetite granules were synthesized by thermal conversion of set brushite cement. Eight female New Zealand rabbits were used for this study. Two identical 10mm diameter bicortical cranial defects were created in each animal. One of the defects was grafted with monetite granules while the contralateral was grafted with granules of intramembranous autologous bone as control. Animals were sacrificed 8 weeks after surgery and biopsies were taken for histological and histomorphometrical evaluation under light microscopy. Wilcoxon test was used for statistical analysis. The bone defects treated with either autologous bone or monetite granules were able to heal within the study period. Upon histological observation the defects treated with autologous bone granules resembled the adjacent intact calvaria, whereas the defects treated with monetite showed a high infiltration of new bone and only 13.4±8.4% of remaining granules. The percentage of bone volume in the defects of the control group (71±9%) was 16% higher than in the study group (55±10%) (p<0.05). The percentage of augmented mineralized tissue volume in the study group (68±18%) was not significantly different from the control group (p>0.05). The amount of augmented mineralized tissue in the bone defects obtained with monetite granules was not significantly different from that obtained with autologous bone. This study confirms the potential of monetite based biomaterials as an alternative to autologous bone graft.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
25190
4-Chloro-2-methylphenoxyacetic acid, technical, ≥95.0% (T)
C9H9ClO3
C4830
Calcium carbonate, BioXtra, ≥99.0%
CCaO3
C5929
Calcium carbonate, BioReagent, suitable for insect cell culture, ≥99.0%
CCaO3
481807
Calcium carbonate, 99.999% trace metals basis
CCaO3
202932
Calcium carbonate, ≥99.995% trace metals basis
CCaO3
398101
Calcium carbonate, ACS reagent, chelometric standard, 99.95-100.05% dry basis
CCaO3
310034
Calcium carbonate, powder, ≤50 μm particle size, 98%
CCaO3
21061
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
CCaO3
C7263
Calcium phosphate dibasic, 98.0-105.0%
HCaO4P
C8606
Dibasic Calcium Phosphate, meets USP testing specifications
HCaO4P
454338
Dicyclopentadiene, contains BHT as stabilizer
C10H12
P5811
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
H3O4P
466123
Phosphoric acid, crystalline, ≥99.999% trace metals basis
H3O4P
452289
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
H3O4P
345245
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
H3O4P
79617
Phosphoric acid, BioUltra, ≥85% (T)
H3O4P
40266
Phosphoric acid, semiconductor grade PURANAL (Honeywell 17861), ≥85%
H3O4P
612162
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 4.2 mm × 8 in. , WGS-5BL Coaxial NMR tube
H3O4P
699608
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 3 mm × 8 in.
H3O4P
698822
Phosphoric acid solution, NMR reference standard, 85% in D2O (99.9 atom % D), NMR tube size 5 mm × 8 in.
H3O4P
W290017
Phosphoric acid solution, 85 wt. % in H2O, FCC, FG
H3O4P
P8010
Sodium pyrophosphate tetrabasic, ≥95%
Na4O7P2