EMAIL THIS PAGE TO A FRIEND
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences
Lactate dehydrogenase inhibitors sensitize lymphoma cells to cisplatin without enhancing the drug effects on immortalized normal lymphocytes.
PMID 25930121
Abstract
Up-regulation of glycolysis, a well recognized hallmark of cancer cells, was also found to be predictive of poor chemotherapy response. This observation suggested the attempt of sensitizing cancer cells to conventional chemotherapeutic agents by inhibiting glucose metabolism. Lactate dehydrogenase (LDH) inhibition can be a way to hinder glycolysis of cancer cells without affecting the metabolism of normal tissues, which usually does not require this enzymatic activity. In this paper, we showed that two LDH inhibitors (oxamate and galloflavin) can increase the efficacy of cisplatin in cultured Burkitt's lymphoma (BL) cells and that this potentiating effect is not exerted in proliferating normal lymphocytes. This result was explained by the finding that in BL cells LDH inhibition induced reactive oxygen species (ROS) generation, which was not evidenced in proliferating normal lymphocytes. In BL cells treated with the association of cisplatin and LDH inhibitors, these ROS can be a further cause of DNA damage, to be added to that produced by cisplatin, leading to the failure of the response repair. At present LDH inhibitors suitable for clinical use are actively searched; our results can allow a better understanding of the potentiality of LDH as a possible target to develop innovative anticancer treatments.