EMAIL THIS PAGE TO A FRIEND
Clinical immunology (Orlando, Fla.)
Differential expression of CD57 in antigen-reactive CD4+ T cells between active and latent tuberculosis infection.
PMID 25931385
Abstract
The development of diagnostic tests that predict the progression of latent tuberculosis infection to active disease is pivotal for the eradication of tuberculosis. As an initial step to achieve this goal, our study's aim was to identify biomarkers that differentiate active from latent tuberculosis infection. We compared active and latent tuberculosis infection groups in terms of the precursor frequency, functional subset differentiation, and senescence/exhaustion surface marker expression of antigen-specific CD4(+) T cells, which were defined as dividing cells upon their encountering with Mycobacterium (M.) tuberculosis antigens. Among several parameters shown to have statistically significant differences between the two groups, the frequency of CD57-expressing cells could differentiate effectively between active disease and latent infection. Our results suggest that the expression of CD57 in M. tuberculosis-reactive CD4(+) T cells could be a promising candidate biomarker with which to identify individuals with latent tuberculosis infection prone to progression to active disease.