EMAIL THIS PAGE TO A FRIEND
Journal of the neurological sciences
Adult brainstem gliomas: Correlation of clinical and molecular features.
PMID 25934342
Abstract
Brainstem gliomas are rare in adults and overall have superior survival outcomes compared to pediatric brainstem gliomas. We conducted a retrospective data and tissue analysis of all adult patients (≥ 18 years old) with World Health Organization (WHO) Grade II, III, and IV brainstem gliomas in the University of Texas MD Anderson Cancer Center institutional database from 1990 to 2012. We identified 143 cases in adults ages 18 and over. There were 28 glioblastomas, 43 anaplastic astrocytomas, 15 diffuse astrocytomas, and 11 gliomas not otherwise specified, and in 46 cases the diagnosis was made radiographically. 128 (89.5%) cases were classified radiographically as diffuse and of the focal tumors, 9 of the 15 were WHO Grade III or IV tumors. Increasing tumor grade and contrast enhancement were associated with significantly reduced overall survival. The median overall survival for the entire cohort was 32.1 months similar to previously published studies. Two of 25 grade II and III tumors, and 1 of 17 glioblastomas had IDH1 mutations on immunohistochemical testing. Nine cases had sufficient tissue for mutation profiling, 1 case had a BRAF V600E mutation and 2 had 2 PIK3CA mutations. Survival outcomes for adult WHO Grade II to IV brainstem gliomas were similar to supratentorial IDH1 wild-type tumors of similar grade and histology. Potentially actionable mutations can be identified from small biopsy samples in a subset of adult brainstem gliomas.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
73248
Melengestrol acetate, ≥97.0% (HPLC)
C25H32O4