EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Heat shock factor 1 prevents the reduction in thrashing due to heat shock in Caenorhabditis elegans.
PMID 25935486
Abstract
Heat shock factor 1 (HSF-1) is activated by heat stress and induces the expression of heat shock proteins. However, the role of HSF-1 in thermotolerance remains unclear. We previously reported that heat stress reversibly reduces thrashing movement in Caenorhabditis elegans. In this study, we analyzed the function of HSF-1 on thermotolerance by monitoring thrashing movement. hsf-1 RNAi suppressed the restoration of thrashing reduced by heat stress. In contrast, hsf-1 knockdown cancelled prevention of movement reduction in insulin/IGF-1-like growth factor 1 receptor (daf-2) mutant, but didn't suppress thrashing restoration in daf-2 mutant. In addition, hsf-1 RNAi accelerated the reduction of thrashing in heat-shocked wild-type C. elegans. And, daf-16 KO didn't accelerate the reduction of thrashing by heat stress. Taken together, these results suggest that HSF-1 prevents the reduction of thrashing caused by heat shock.