EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Bioactivity assay of porcine relaxin based on cAMP accumulation in THP-1 cells quantified by LC-MS/MS.
PMID 25935614
Abstract
This study describes a reliable bioactivity assay of porcine relaxin (pRLX) based on cyclic adenosine 3',5'-monophosphate (cAMP) accumulation in the human monocyte cell-line quantified by liquid chromatography-tandem mass spectrometry (LC-MS/MS). As a result, the LC-MS/MS was based on a positive selected reaction monitoring of cAMP with a stable internal standard, 8-Br-cAMP and a protein precipitation procedure by HClO4. The standard curve of cAMP was linear from 5.0 ng mL(-1) to 992.0 ng mL(-1), with lower limits of detection and quantification of 0.5 ng mL(-1) and 5.0 ng mL(-1), respectively. The satisfactory validation data including stability assay were obtained. When measured by the LC-MS/MS, the pRLX sample showed a time- and dose-dependent stimulation of cAMP with the concentration for 50% of the maximal effect (EC50) of 40.6 ng mL(-1). The developed method indicated higher precision and selectivity than the commercial enzyme linked immunosorbent assay kits, which showed EC50 of 66.6 ng mL(-1).