EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Electrochemical vapor deposition of semiconductors from gas phase with a solid membrane cell.
PMID 25936392
Abstract
We demonstrate the feasibility of semiconductor deposition via the electrochemical reduction of gaseous precursors by the use of an anhydrous proton-conducting membrane, the solid acid CsHSO4, at 165 °C. This membrane electrode assembly was operated within the oxidation of hydrogen on a porous Pt anode and the deposition of Si or Ge under bias at the cathode from chloride-based gaseous precursors; SiCl4 and GeCl4 in an Ar flow with a reduction potential over -1.0 V (vs RHE).