PloS one
Interobserver Agreement between On-Call Radiology Resident and General Radiologist Interpretations of CT Pulmonary Angiograms and CT Venograms.
PMID 25938666
Abstract
To evaluate the interobserver agreement (IOA) between the initial radiology resident and the final staff radiologist reports of combined computed tomographic pulmonary angiograms (CTPA) and computed tomographic venograms (CTV) performed during on-call hours. Approval by the institutional review board was obtained. Six-hundred and ninety-six consecutive studies (CTPA or CTPA with CTV) performed during on-call hours and interpreted by 30 residents were identified. Radiology residents' reports were compared to the final staff reports. Three tests outcomes were considered (positive, P; negative, N; indeterminate, I). Discordant cases were reviews by a chest radiologist. CTPAs were reported by staff radiologists as positive for pulmonary embolism (PE) in 18% (126/694), with a kappa of 0.81 (95% CI 0.77-0.86) with 3 outcomes (P, N, I), and a kappa of 0.89 (95% CI 0.85-0.94) with 2 outcomes (P, N). Regarding PE location, good concordance was observed for positive studies, with a kappa of 0.86 (95% CI 0.78 - 0.95). CTVs were reported as positive by staff radiologists in 8.5% (33/388), with a kappa of 0.66 (95% CI 0.55-0.77) with 3 outcomes (P, N, I), and a kappa of 0.89 (95% CI 0.8-1.0) with 2 outcomes (P, N). The IOA between residents and staff radiologists increased with increasing residency year level for CTPAs, but did not for CTVs. Very good and good IOA were observed between resident and staff radiologist interpretations for CTPA and CTV, respectively, with tendency towards improved IOA as residency level of training increased for CTPA, but not for CTV.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D2158
Histodenz, nonionic density gradient medium
C19H26I3N3O9