EMAIL THIS PAGE TO A FRIEND
BMC neuroscience
Increase in the titer of lentiviral vectors expressing potassium channels by current blockade during viral vector production.
PMID 25940378
Abstract
High titers of lentiviral vectors are required for the efficient transduction of a gene of interest. During preparation of lentiviral the vectors, the protein of interest is inevitably expressed in the viral vector-producing cells. This expression may affect the production of the lentiviral vector. We prepared lentiviral vectors expressing inwardly rectifying potassium channel (Lv-Kir2.1), its dominant-negative form (Lv-Kir-DN), and other K(+) channels, using the ubiquitously active β-actin and neuron-specific synapsin I promoters. The titer of Lv-Kir-DN was higher than that of Lv-Kir2.1, suggesting a negative effect of induced K(+) currents on viral titer. We then blocked Kir2.1 currents with the selective blocker Ba(2+) during Lv-Kir2.1 production, and obtained about a 5-fold increase in the titer. Higher extracellular K(+) concentrations increased the titer of Lv-Kir2.1 about 9-fold. With a synapsin I promoter Ba(2+) increased the titer because of the moderate expression of Kir2.1 channel. Channel blockade also increased the titers of the lentivirus expressing Kv1.4 and TREK channels, but not HERG. The increase in titer correlated with the K(+) currents generated by the channels expressed. In the production of lentivirus expressing K(+) channels, titers are increased by blocking K(+) currents in the virus-producing cells. This identifies a crucial issue in the production of viruses expressing membrane channels, and should facilitate basic and gene therapeutic research on channelopathies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
MAB377
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse