EMAIL THIS PAGE TO A FRIEND
Small (Weinheim an der Bergstrasse, Germany)
pH-Dependent Galvanic Replacement of Supported and Colloidal Cu2O Nanocrystals with Gold and Palladium.
PMID 25940945
Abstract
Galvanic replacement reactions (GRRs) on nanoparticles (NPs) are typically performed between two metals, i.e., a solid metal NP and a replacing salt solution of a more noble metal. The solution pH in GRRs is commonly considered an irrelevant parameter. Yet, the solution pH plays a major role in GRRs involving metal oxide NPs. Here, Cu(2)O nanocrystals (NCs) are studied as galvanic replacement (GR) precursors, undergoing replacement by gold and palladium, with the resulting nanostructures showing a strong dependence on the pH of the replacing metal salt solution. GRRs are reported for the first time on supported (chemically deposited) oxide NCs and the results are compared with those obtained with corresponding colloidal systems. Control of the pH enables production of different nanostructures, from metal-decorated Cu(2)O NCs to uniformly coated Cu(2)O-in-metal (Cu(2)O@Me) core-shell nanoarchitectures. Improved metal nucleation efficiencies at low pHs are attributed to changes in the Cu(2)O surface charge resulting from protonation of the oxide surface. GR followed by etching of the Cu(2)O cores provides metal nanocages that collapse upon drying; the latter is prevented using a sol-gel silica overlayer stabilizing the metal nanocages. Metal-replaced Cu(2)O NCs and their corresponding stabilized nanostructures may be useful as photocatalysts, electrocatalysts, and nanosensors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A3648
(3-Aminopropyl)triethoxysilane, ≥98%
C9H23NO3Si
706493
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
C9H23NO3Si
741442
(3-Aminopropyl)triethoxysilane, ≥98.0%
C9H23NO3Si
281778
(3-Aminopropyl)trimethoxysilane, 97%
C6H17NO3Si
A7222
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, ≥96.0% (HPCE), solid
C16H8NNa3O9S3
09341
8-Aminopyrene-1,3,6-trisulfonic acid trisodium salt, BioReagent, suitable for fluorescence, ≥96.0% (HPCE)
C16H8NNa3O9S3
651745
Copper(I) chloride, anhydrous, beads, ≥99.99% trace metals basis
ClCu
229628
Copper(I) chloride, ≥99.995% trace metals basis
ClCu
225819
Hydrazine hydrate, reagent grade, N2H4 50-60 %
H4N2 · xH2O
207942
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, 98%
H4N2 · H2O
W302600
Sodium citrate dihydrate, ≥99%, FG
C6H5Na3O7 · 2H2O
S4641
Sodium citrate tribasic dihydrate, ACS reagent, ≥99.0%
C6H5Na3O7 · 2H2O
C7254
Sodium citrate tribasic dihydrate, ≥98%
C6H5Na3O7 · 2H2O
C8532
Sodium citrate tribasic dihydrate, for molecular biology, ≥99%
C6H5Na3O7 · 2H2O
C3434
Sodium citrate tribasic dihydrate, BioXtra, ≥99.0% (titration)
C6H5Na3O7 · 2H2O
71402
Sodium citrate tribasic dihydrate, BioUltra, for molecular biology, ≥99.5% (NT)
C6H5Na3O7 · 2H2O
71405
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)
C6H5Na3O7 · 2H2O
S1804
Trisodium citrate dihydrate, meets USP testing specifications
C6H5Na3O7 · 2H2O