EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
De novo-developed antibodies to donor MHC antigens lead to dysregulation of microRNAs and induction of MHC class II.
PMID 25941328
Abstract
Immune responses to HLA and development of anti-donor HLA (DSA) were shown to play a role in chronic rejection following transplantation. We hypothesized that Abs to MHC change microRNAs (miRNAs), leading to chronic lung allograft rejection. Microarray analysis was performed in a murine model of anti-MHC-induced obliterative airway disease (OAD), a correlate of obliterative bronchiolitis. A unique profile of dysregulated miRNAs was detected in OAD mice on days 7 and 15 after Ab administration compared with control. Sixty-seven miRNAs were increased and 42 miRNAs were decreased in OAD mice on day 7. In addition, 15 miRNAs were overexpressed and 16 miRNAs were underexpressed in OAD mice on day 15. The expression of miR-16 and miR-195 was significantly decreased in lungs of OAD mice, as assessed by quantitative RT-PCR and in situ hybridization, with increases in H-2 Aa and H-2 Dma mRNA levels. Significant reductions in miR-16 and miR-195 levels were also noted in lung transplant (LTx) patients with DSA compared with LTx patients without DSA. Bioinformatic TargetScan and reporter assays identified the binding of miR-16 and miR-195 to the 3'-untranslated region of regulatory factor X 5. Quantitative PCR and immunohistochemistry indicated posttranscriptional increases in regulatory factor X 5 mRNA and protein expression in OAD mice, as well as in LTx recipients with DSA, which was associated with increased expression of HLA-DPA1, HLA-DQA1, and HLA-DRA mRNA. Therefore, our results demonstrated that miRNAs induced by alloimmunity may play important roles in chronic rejection after LTx.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D6770
Digoxin, analytical standard
C41H64O14