EMAIL THIS PAGE TO A FRIEND
Oncoimmunology
Heteroclitic XBP1 peptides evoke tumor-specific memory cytotoxic T lymphocytes against breast cancer, colon cancer, and pancreatic cancer cells.
PMID 25941601
Abstract
XBP1 is a critical transcriptional activator of the unfolded protein response (UPR), which increases tumor cell survival under prolonged endoplasmic reticulum (ER) stress and hypoxic conditions.This study was designed to evaluate the immunogenicity of heteroclitic XBP1 unspliced (US)