EMAIL THIS PAGE TO A FRIEND
Molecular nutrition & food research
Resveratrol improves hepatic steatosis by inducing autophagy through the cAMP signaling pathway.
PMID 25943029
Abstract
Resveratrol (RSV), a natural polyphenol, has been reported to attenuate nonalcoholic fatty liver disease (NAFLD); however, its underlying mechanism is unclear. Autophagy was recently identified as a critical protective mechanism during NAFLD development. Therefore, we investigated the role of autophagy in the beneficial effects of RSV on hepatic steatosis. Via Oil red O staining, triglyceride, and β-hydroxybutyrate detection, we found that RSV decreased palmitate-induced lipid accumulation and stimulated fatty acid β-oxidation in hepatocytes. Based on Western blot assay, confocal microscopy and transmission electron microscopy, we found that RSV induced autophagy in hepatocytes, whereas autophagy inhibition markedly abolished RSV-mediated hepatic steatosis improvement. Moreover, RSV increased cAMP levels and the levels of SIRT1 (sirtuin 1), pPRKA (phosphorylated protein kinase A), and pAMPK (phosphorylated AMP-activated protein kinase), as well as SIRT1 activity in HepG2 cells. Incubation with inhibitors of AC (adenylyl cyclase), PRKA, AMPK, SIRT1, or with AC, PRKA, AMPK, or SIRT1 siRNA abolished RSV-mediated autophagy. Similar results were obtained in mice with hepatic steatosis. RSV improved hepatic steatosis partially by inducing autophagy via the cAMP-PRKA-AMPK-SIRT1 signaling pathway, which provides new evidence regarding RSV's effects on NAFLD treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
K3394
KH7, ≥98% (HPLC)
C17H15BrN4O2S