EMAIL THIS PAGE TO A FRIEND
Nature communications
Prdx4 is a compartment-specific H2O2 sensor that regulates neurogenesis by controlling surface expression of GDE2.
PMID 25943695
Abstract
Neural progenitors and terminally differentiated neurons show distinct redox profiles, suggesting that coupled-redox cascades regulate the initiation and progression of neuronal differentiation. Discrete cellular compartments have different redox environments and how they contribute to differentiation is unclear. Here we show that Prdx4, an endoplasmic reticulum (ER) enzyme that metabolizes H2O2, acts as a tunable regulator of neurogenesis via its compartmentalized thiol-oxidative function. Prdx4 ablation causes premature motor neuron differentiation and progenitor depletion, leading to imbalances in subtype-specific motor neurons. GDE2, a six-transmembrane protein that induces differentiation by downregulating Notch signalling through surface cleavage of GPI-anchored proteins, is targeted by Prdx4 oxidative activity. Prdx4 dimers generated by H2O2 metabolism oxidize two cysteine residues within the GDE2 enzymatic domain, which blocks GDE2 trafficking to the plasma membrane and prevents GDE2 neurogeneic function. Thus, Prdx4 oxidative activity acts as a sensor to directly couple neuronal differentiation with redox environments in the ER.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
691275
Acetic anhydride, Lonza quality, ≥99.5% (GC)
C4H6O3
242845
Acetic anhydride, ACS reagent, ≥98.0%
C4H6O3
320102
Acetic anhydride, ReagentPlus®, ≥99%
C4H6O3
539996
Acetic anhydride, 99.5%
C4H6O3
A6404
Acetic anhydride, reagent grade, ≥98%
C4H6O3
01-0390
Acetic anhydride, SAJ first grade, ≥93.0%
C4H6O3
01-0380
Acetic anhydride, JIS special grade
C4H6O3
L095004
Acetic anhydride
C4H6O3
MAB1501
Anti-Actin Antibody, clone C4, Reliably and specifically detect actin using this Anti-Actin Antibody, clone C4. This highly published monoclonal antibody is validated for use in ELISA, IC, IF, IH, IH(P) & WB. This mAb is also available as a fluorescent conjugate.
05-368MG
Anti-phospho-Ser/Thr-Pro MPM-2 Antibody, Detect phospho-Ser/Thr-Pro MPM-2 also known as Mitotic protein #2 with Anti-phospho-Ser/Thr-Pro MPM-2 Antibody (Mouse Monoclonal Antibody), that has been demonstrated to work in ELISA, IHC, IP & WB.
F9037
Formamide, ≥99.5% (GC), BioReagent, for molecular biology
CH3NO
F7508
Formamide, for molecular biology, ≥99.5% (GC), liquid
CH3NO
221198
Formamide, ACS reagent, ≥99.5%
CH3NO
47671
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
CH3NO
185906
Formamide, reagent grade, 98%
CH3NO
733784
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
CH3NO
736201
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
CH3NO
E3876
N-Ethylmaleimide, crystalline, ≥98% (HPLC)
C6H7NO2
E1271
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
C6H7NO2
04259
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
C6H7NO2
R6750 Ribonucleic acid from baker's yeast (S. cerevisiae)
ARK2174
Triethanolamine, PharmaGrade, USP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production.
C6H15NO3
90278
Triethanolamine, BioUltra, ≥99.5% (GC)
C6H15NO3
90279
Triethanolamine, ≥99.0% (GC)
C6H15NO3
33729
Triethanolamine, puriss. p.a., ≥99% (GC)
C6H15NO3
T58300
Triethanolamine, reagent grade, 98%
C6H15NO3
X114
Triton X-114, laboratory grade