EMAIL THIS PAGE TO A FRIEND
Polish journal of pharmacology and pharmacy
Preliminary pharmacological evaluation of chelidonine in rodents.
PMID 2594578
Abstract
Our studies showed that the chelidonine administered intraperitoneally (ip) exerted an inhibitory effect on the central nervous system of the rodents. It was shown that chelidonine depressed the spontaneous and explorative motor activities, decreased body temperature, potentiated the action of hypnotics and increased sedation in reserpinized mice. Chelidonine did not affect the reactivity of the mice in the hot-plate test, and was without any protective effect against pentylenetetrazol-induced seizures. This alkaloid antagonized the head twitch response induced by 5-hydroxytryptophan (5-HTP) or 5-methoxytryptamine (5-MT) in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54274
Chelidonine, ≥97.0% (HPLC)
C20H19NO5