EMAIL THIS PAGE TO A FRIEND
Autophagy
Uvrag targeting by Mir125a and Mir351 modulates autophagy associated with Ewsr1 deficiency.
PMID 25946189
Abstract
The EWSR1 (EWS RNA-binding protein 1/Ewing Sarcoma Break Point Region 1) gene encodes a RNA/DNA binding protein that is ubiquitously expressed and involved in various cellular processes. EWSR1 deficiency leads to impairment of development and accelerated senescence but the mechanism is not known. Herein, we found that EWSR1 modulates the Uvrag (UV radiation resistance associated) gene at the post-transcription level. Interestingly, EWSR1 deficiency led to the activation of the DROSHA-mediated microprocessor complex and increased the level of Mir125a and Mir351, which directly target Uvrag. Moreover, the Mir125a- and Mir351-mediated reduction of Uvrag was associated with the inhibition of autophagy that was confirmed in ewsr1 knockout (KO) MEFs and ewsr1 KO mice. Taken together, our data indicate that EWSR1 is involved in the post-transcriptional regulation of Uvrag via a miRNA-dependent pathway, resulting in the deregulation of autophagy inhibition. The mechanism of Uvrag and autophagy regulation by EWSR1 provides new insights into the role of EWSR1 deficiency-related cellular dysfunction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
94640
DL-Valine, ReagentPlus®, ≥99.0% (NT)
C5H11NO2
W344400
DL-Valine, ≥97%
C5H11NO2
M7145 MEM Non-essential Amino Acid Solution (100×), without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
EMU019231 MISSION® esiRNA, esiRNA targeting mouse Uvrag (esiRNA1)
EHU061641 MISSION® esiRNA, esiRNA human UVRAG (esiRNA1)
P9333
Potassium chloride, BioXtra, ≥99.0%
ClK
P9541
Potassium chloride, for molecular biology, ≥99.0%
ClK
P5405
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
ClK
449989
Potassium chloride, anhydrous, beads, −10 mesh, 99.999% trace metals basis
ClK
451029
Potassium chloride, anhydrous, beads, −10 mesh, 99.99% trace metals basis
ClK
409316
Potassium chloride, 99.999% trace metals basis
ClK
204099
Potassium chloride, ≥99.99% trace metals basis
ClK
60128
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
ClK
P9327
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
ClK
60142
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
ClK
688770
tert-Butyl acetoacetate, Lonza quality, ≥98.0% (w/w) (GC)
C8H14O3
537594
tert-Butyl acetoacetate, reagent grade, 98%
C8H14O3
T9159
Trichloroacetic acid, BioXtra, ≥99.0%
C2HCl3O2
T6399
Trichloroacetic acid, ACS reagent, ≥99.0%
C2HCl3O2
T4885
Trichloroacetic acid, ≥99.0% (titration)
C2HCl3O2
T8657
Trichloroacetic acid, for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
C2HCl3O2
91228
Trichloroacetic acid, BioUltra, ≥99.5% (T)
C2HCl3O2
91230
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
C2HCl3O2
27242
Trichloroacetic acid, puriss., meets analytical specification of Ph. Eur., USP 21, 99-100.5% (calc. to the dried substance)
C2HCl3O2
T0699
Trichloroacetic acid solution, 6.1 N
C2HCl3O2