EMAIL THIS PAGE TO A FRIEND
Mediators of inflammation
Enhanced levels of chemokines and their receptors in the colon of microscopic colitis patients indicate mixed immune cell recruitment.
PMID 25948880
Abstract
Microscopic colitis (MC), comprising collagenous colitis (CC) and lymphocytic colitis (LC), is a common cause of chronic diarrhea. Various immune cell infiltrations in the epithelium and lamina propria are seen in MC immunopathology. We compared gene and protein expressions of different immune cell attracting chemokines and their receptors in colon biopsies from MC patients in active disease or histopathological remission (CC/LC-HR) with controls, using qRT-PCR and Luminex, respectively. CC and LC patients with active disease demonstrated a mixed chemokine profile with significantly enhanced gene and/or protein expressions of the chemokines CCL2, CCL3, CCL4, CCL5, CCL7, CCL22, CXCL8, CXCL9, CXCL10, CXCL11, and CX3CL1 and the receptors CCR2, CCR3, CCR4, CXCR1, CXCR2, and CX3CR1. Enhanced chemokine/chemokine receptor gene and protein levels in LC-HR patients were similar to LC patients, whereas CC-HR patients demonstrated almost normalized levels. These findings expand the current understanding of the involvement of various immune cells in MC immunopathology and endorse chemokines as potential diagnostic markers as well as therapeutic candidates. Moreover, this study further supports the hypothesis that CC and LC are two different entities due to differences in their immunoregulatory responses.