EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Excretory, Secretory, and Tissue Residues after Label and Extra-label Administration of Flunixin Meglumine to Saline- or Lipopolysaccharide-Exposed Dairy Cows.
PMID 25950946
Abstract
Twenty lactating dairy cattle were intravenously infused with either lipopolysaccharide (LPS) (n = 10) or sterile saline (n = 10). Five cattle in each group received three doses of flunixin meglumine administered by either intravenous infusion or intramuscular injection at 24 h intervals. Milk, urine, and tissues were collected. Thirty-six hours after the last flunixin administration, milk from six cows contained 5-hydroxyflunixin (5OHF) levels greater than the milk tolerance of 2 ng/mL; by 48 h, milk from two cows, a saline and a LPS-treated animal, had violative milk concentrations of 5OHF. A single animal treated with LPS and intramuscular flunixin contained violative flunixin residues in liver. The ratio of urinary flunixin/5OHF was correlated (P < 0.01; R(2) = 0.946) with liver flunixin residues in LPS-treated animals, but not (P = 0.96; R(2) = 0.003) in cows treated with saline in lieu of LPS. Violative residues of flunixin in dairy cattle may be related to LPS inhibition of flunixin metabolism.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F0429
Flunixin meglumine, ≥98% (HPLC)
C14H11F3N2O2·C7H17NO5