EMAIL THIS PAGE TO A FRIEND
Journal of cellular physiology
S100A8, An Oocyte-Specific Chemokine, Directs the Migration of Ovarian Somatic Cells During Mouse Primordial Follicle Assembly.
PMID 25953201
Abstract
In the mammalian ovaries, the primordial follicle pool determines the reproductive capability over the lifetime of a female. The primordial follicle is composed of two cell members, namely the oocyte and the pre-granulosa cells that encircle the oocyte. However, it is unclear what factors are involved in the reorganization of the two distinct cells into one functional unit. This study was performed to address this issue. Firstly, in an in vitro reconstruction system, dispersed ovarian cells from murine fetal ovaries at 19.0 days post coitum (dpc) reassembled into follicle-like structures, independent of the physical distance between the cells, implying that either oocytes or ovarian somatic cells (OSCs) were motile. We then carried out a series of transwell assay experiments, and determined that it was in fact 19.0 dpc OSCs (as opposed to oocytes), which exhibited a significant chemotactic response to both fetal bovine serum and oocytes themselves. We observed that S100A8, a multi-functional chemokine, may participate in the process as it is mainly expressed in oocytes within the cysts/plasmodia. S100A8 significantly promoted the number of migrating OSCs by 2.5 times in vitro, of which 66.9% were FOXL2 protein-positive cells, implying that the majority of motile OSCs were pre-granulosa cells. In addition, an S100A8-specific antibody inhibited the formation of follicle-like reconstruction cell mass in vitro. And, the primordial follicle formation was reduced when S100a8-specific siRNA was applied onto in vitro cultured 17.5 dpc ovary. Therefore, S100A8 could be a chemokine of oocyte origin, which attracts OSCs to form the primordial follicles.