EMAIL THIS PAGE TO A FRIEND
Frontiers in Cellular Neuroscience
Retinal aging in the diurnal Chilean rodent (Octodon degus): histological, ultrastructural and neurochemical alterations of the vertical information processing pathway.
PMID 25954153