EMAIL THIS PAGE TO A FRIEND
Diseases of aquatic organisms
Testing of candidate non-lethal sampling methods for detection of Renibacterium salmoninarum in juvenile Chinook salmon Oncorhynchus tshawytscha.
PMID 25958804
Abstract
Non-lethal pathogen testing can be a useful tool for fish disease research and management. Our research objectives were to determine if (1) fin clips, gill snips, surface mucus scrapings, blood draws, or kidney biopsies could be obtained non-lethally from 3 to 15 g Chinook salmon Oncorhynchus tshawytscha, (2) non-lethal samples could accurately discriminate between fish exposed to the bacterial kidney disease agent Renibacterium salmoninarum and non-exposed fish, and (3) non-lethal samples could serve as proxies for lethal kidney samples to assess infection intensity. Blood draws and kidney biopsies caused ≥5% post-sampling mortality (Objective 1) and may be appropriate only for larger fish, but the other sample types were non-lethal. Sampling was performed over 21 wk following R. salmoninarum immersion challenge of fish from 2 stocks (Objectives 2 and 3), and nested PCR (nPCR) and real-time quantitative PCR (qPCR) results from candidate non-lethal samples were compared with kidney tissue analysis by nPCR, qPCR, bacteriological culture, enzyme-linked immunosorbent assay (ELISA), fluorescent antibody test (FAT) and histopathology/immunohistochemistry. R. salmoninarum was detected by PCR in >50% of fin, gill, and mucus samples from challenged fish. Mucus qPCR was the only non-lethal assay exhibiting both diagnostic sensitivity and specificity estimates>90% for distinguishing between R. salmoninarum-exposed and non-exposed fish and was the best candidate for use as an alternative to lethal kidney sample testing. Mucus qPCR R. salmoninarum quantity estimates reflected changes in kidney bacterial load estimates, as evidenced by significant positive correlations with kidney R. salmoninarum infection intensity scores at all sample times and in both fish stocks, and were not significantly impacted by environmental R. salmoninarum concentrations.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N7000
1-Naphthyl phosphate monosodium salt monohydrate, ≥98% (titration), powder
C10H8NaO4P · H2O
C9157
Activated charcoal, suitable for cell culture, suitable for plant cell culture
C
440345
Glass spheres
O2Si
420875
LUDOX® AM colloidal silica, 30 wt. % suspension in H2O
O2Si
420832
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420840
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420883
LUDOX® CL colloidal silica, 30 wt. % suspension in H2O
O2Si
420891
LUDOX® CL-X colloidal silica, 45 wt. % suspension in H2O
O2Si
420824
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
O2Si
420816
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420808
LUDOX® LS colloidal silica, 30 wt. % suspension in H2O
O2Si
420794
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
O2Si
420786
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
O2Si
420778
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
O2Si
420859
LUDOX® TMA colloidal silica, 34 wt. % suspension in H2O
O2Si
204358
Sand, white quartz, ≥99.995% trace metals basis
O2Si
748161
Silica, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
O2Si
718483
Silica, nanopowder, spec. surface area 175-225 m2/g (BET), 99.8% trace metals basis
O2Si
381276
Silica, 99.8%
O2Si
421553
Silica
O2Si
S5130
Silica, fumed, powder
O2Si
S5505
Silica, fumed, powder, 0.2-0.3 μm avg. part. size (aggregate)
O2Si
643645
Silica, mesostructured, MCM-41 type (hexagonal)
O2Si
777242
Silica, mesostructured, SBA-15, 99% trace metals basis
O2Si
560979
Silica, mesostructured, MSU-F (cellular foam)
O2Si
541036
Silica, mesostructured, HMS (wormhole)
O2Si
637238
Silicon dioxide, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
O2Si
637246
Silicon dioxide, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
O2Si
342831
Silicon dioxide, fused (granular), 4-20 mesh, 99.9% trace metals basis
O2Si
342890
Silicon dioxide, −325 mesh, 99.5% trace metals basis
O2Si