EMAIL THIS PAGE TO A FRIEND
Toxicon : official journal of the International Society on Toxinology
Prothrombin activator-like toxin appears to mediate cardiovascular collapse following envenoming by Pseudonaja textilis.
PMID 25959508
Abstract
Brown snake (Pseudonaja spp.)-induced early cardiovascular collapse is a life-threatening medical emergency in Australia. We have previously shown that this effect can be mimicked in animals and is mediated via the release of endogenous mediators. In the present study, we aimed to purify and characterize the component in Pseudonaja textilis venom which induces cardiovascular collapse following envenoming. The component (fraction 3) was isolated using a combination of techniques including hydroxyapatite and reverse phase chromatography. Fraction 3 (10 or 20xa0μg/kg, i.v.) produced a rapid decrease in mean arterial pressure (MAP) followed by cardiovascular collapse. Fraction 3-induced early collapse was abolished by prior administration of smaller priming doses of fraction 3 (i.e. 2 and 5xa0μg/kg, i.v.) or heparin (300 units/kg, i.v.). P.xa0textilis whole venom (1 and 3xa0μg/ml), but not fraction 3 (1 or 3xa0μg/ml), induced endothelium-dependent relaxation in isolated rat mesenteric arteries. SDS-PAGE gel indicated the presence of 9-10 protein bands of fraction 3. Using proteomic based analysis some protein bands of fraction 3 were identified as subunits of venom prothrombin activator, pseutarin C of P.xa0textilis venom. Our results conclude that prothrombin activator-like toxin is likely to be a contributor to the rapid collapse induced by P.xa0textilis venom.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P5514
Pyrilamine maleate salt
C17H23N3O · C4H4O4