EMAIL THIS PAGE TO A FRIEND
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society
Enteric glial cells are associated with stress-induced colonic hyper-contraction in maternally separated rats.
PMID 25960044
Abstract
Enteric glial cells (EGCs) play important roles in enteric integrity and regulation of gastrointestinal function. However, whether EGCs undergo pathophysiological changes in stress-associated gastrointestinal disorders is unknown. We investigated structural and functional alterations in colonic EGCs and their roles in colonic contraction in an irritable bowel syndrome (IBS) model. As a chronic stress, male Wistar rats underwent 3-h maternal separation during postnatal days 2-14. As an acute stress, we used water-immersion stress (4 h) in adulthood (at 8 weeks). We quantitatively and morphologically evaluated enteric neurons and EGCs using whole-mount longitudinal muscle-myenteric plexus preparations. Colonic contraction was analyzed with electrical field stimulation (EFS). Glial fibrillary acidic protein (GFAP) expression and the number of total, cholinergic, and nitrergic neurons were unchanged in maternally separated rats with acute stress (combined stress: an IBS model) compared with controls. However, the density of GFAP-positive EGC processes that apparently overlapped with the neurons and the extent of bulbous swelling of terminals increased according to the stress intensity: control, acute stress, maternal separation, and combined stress. EFS-induced colonic contractions were significantly greater in the combined stress rats than in controls. Higher dose of fluorocitrate, a selective inhibitor of EGC metabolism, was required to inhibit both EFS-induced contraction and EGCs activation in the combined stress rats than in controls. Colonic EGCs exhibited structural alterations according to the stress intensity. EGCs were associated with stress-induced colonic hyper-contraction in the combined stress rats, which may underlie the pathogenesis of IBS.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9634
DL-Fluorocitric acid barium salt
C12H8Ba3F2O14