EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Stable and efficient hole transporting materials with a dimethylfluorenylamino moiety for perovskite solar cells.
PMID 25960182
Abstract
Novel star-shaped hole transporting materials (HTMs) with a bis-dimethylfluorenylamino moiety have been synthesized and evaluated for high performance perovskite solar cell applications. Maximum power conversion efficiency of 14.21% has been achieved by using the HTM with a fused TPA core and the long-term stability was also shown to be comparable with that of .
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
284513
Chlorobenzene, anhydrous, 99.8%
C6H5Cl