PloS one
MiR-144 Inhibits Uveal Melanoma Cell Proliferation and Invasion by Regulating c-Met Expression.
PMID 25961751
Abstract
MicroRNAs (miRNAs) are a group endogenous small non-coding RNAs that inhibit protein translation through binding to specific target mRNAs. Recent studies have demonstrated that miRNAs are implicated in the development of cancer. However, the role of miR-144 in uveal melanoma metastasis remains largely unknown. MiR-144 was downregulated in both uveal melanoma cells and tissues. Transfection of miR-144 mimic into uveal melanoma cells led to a decrease in cell growth and invasion. After identification of two putative miR-144 binding sites within the 3' UTR of the human c-Met mRNA, miR-144 was proved to inhibit the luciferase activity inMUM-2B cells with a luciferase reporter construct containing the binding sites. In addition, the expression of c-Met protein was inhibited by miR-144. Furthermore, c-Met-mediated cell proliferation and invasion were inhibited by restoration of miR-144 in uveal melanoma cells. In conclusion, miR-144 acts as a tumor suppressor in uveal melanoma, through inhibiting cell proliferation and migration. miR-144 might serve as a potential therapeutic target in uveal melanoma patients.