EMAIL THIS PAGE TO A FRIEND
BioFactors (Oxford, England)
Taurine, a major amino acid of oyster, enhances linear bone growth in a mouse model of protein malnutrition.
PMID 25963419
Abstract
Oysters (Oys) contain various beneficial components, such as, antioxidants and amino acids. However, the effects of Oys or taurine (Tau), a major amino acid in Oys on bone growth have not been determined. In the present study, we evaluated the effects of Oys or Tau on linear bone growth in a mouse model of protein malnutrition. To make the protein malnutrition in a mouse, we used a low protein diet. Growth plate thickness was increased by Oys or Tau. Bone volume/tissue volume, trabecular thickness, trabecular number, connection density, and total porosity were also improved by Oys or Tau. Oys or Tau increased insulin-like growth factor-1 (IGF-1) levels in serum, liver, and tibia-growth plate. Phosphorylations of Janus kinase 2 (JAK2) and signal transducer and activator of transcription 5 (STAT5) were increased by Oys and by Tau. These findings show that Oys or Tau may increase growth plate thickness by elevating IGF-1 levels and by promoting the phosphorylations of JAK2-STAT5, and suggest that Oys or Tau are growth-promoting substances of potential use in the food and pharmaceutical industries.