EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Characterization of lysozyme adsorption in cellulosic chromatographic materials using small-angle neutron scattering.
PMID 25964231
Abstract
Measurements of the nanoscale structure of chromatographic adsorbents and the associated distribution of sorbed protein within the media can facilitate improvements in such media. We demonstrate a new technique for this purpose using small-angle neutron scattering (SANS) to characterize the nano- to microscale structure of the chromatographic media and sorbed protein under conditions relevant for preparative chromatographic separations. The adsorption of lysozyme on cellulosic S HyperCel™ (Pall Corporation), a strong cation exchanger, was investigated by SANS. The scattering spectrum is reduced to three contributions arising from (1) the chromatographic medium, (2) discrete protein molecules, and (3) the distribution of sorbed protein within the medium. These contributions are quantified for a range of protein loadings. The total concentration of protein in the chromatographic media can be quantified from the SANS spectrum and the protein is observed to retain its tertiary structure upon adsorption, within the resolution of the method. Further analysis of the SANS spectra shows that protein adsorption is uniform in the media. These measurement techniques provide new and valuable nanoscale information about protein sorption in chromatographic media.