EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Study of chemical bonding, physical and biological effect of metformin drug as an organized medicine for diabetes patients with chromium(III) and vanadium(IV) ions.
PMID 25965516
Abstract
New vanadium(IV) and chromium(III) complexes of metformin (MFN) were synthesized upon the chemical interaction between vanadyl(II) sulfate monohydrate or chromium(III) chloride hexahydrate with metformin diabetic drug in the media of a pure grade of methanol solvent. The [(VO)2(MFN)2(SO4)2]2H2O and [Cr(MFN)3]·Cl3·6H2O complexes were discussed using microanalytical measurements, molar conductance, spectroscopic (infrared, ESR, XRD, and UV-vis), effective magnetic moment, scanning electron microscopy (SEM), and thermal analyses (TG/DTG). The elemental analysis shows that VO(II) and Cr(III) complexes were associated with 1:1 and 1:3M ratios, respectively. The infrared spectroscopic results data received from the comparison between free MFN free ligand and their vanadyl(II) and chromium(III) complexes were proven that metformin reacted with respected metal ions as a bidentate ligand through its two imino groups. The kinetic thermodynamic parameters were estimated from the DTG curves. The microstructure changes of the VO(II) and Cr(III) complexes have been probed using positron annihilation lifetime (PAL) and positron annihilation Doppler broadening (PADB) techniques. The PAL and PADB line-shape parameters were found to be dependent on the structure, electronic configuration and molecular weight of metal complexes. Antimicrobial activity of the metformin free ligand and its vanadyl(II) and chromium(III) complexes were evaluated against the gram negative and gram positive bacteria strains and different fungal strains. Moderate antimicrobial activity recorded by disk diffusion inhibition growth zone method in vanadyl(II) and chromium(III) complexes compared to metformin free ligand.