EMAIL THIS PAGE TO A FRIEND
Oncotarget
MicroRNA-155 promotes bladder cancer growth by repressing the tumor suppressor DMTF1.
PMID 25965824
Abstract
MicroRNA-155 (miR-155) is dysregulated in human cancers. In this study, we reported that miR-155 was over-expressed in bladder cancer tissues. We found that miR-155 promoted cell proliferation in vitro and tumorigenesis in vivo. MiR-155 directly reduced the expression of the tumor suppressor DMTF1. The expression of DMTF1 was decreased in bladder cancer tissues. Similar to the restoring miR-155 expression, knockdown of DMTF1 promoted cell growth and cell cycle progression, whereas DMTF1 over-expression rescued the effect of miR-155. Moreover, we investigated DMTF1-Arf-p53 pathway and found that DMTF1 worked in both p53-dependent and p53-independent manners. Taken together, our findings suggested that miR-155 functions as a tumor promoter in bladder cancer, which is partially through repressing DMTF1 expression. The identification of miR-155 and its novel target DMTF1 will be valuable in developing diagnostic markers and therapeutic applications for bladder cancer.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
EMU173171 MISSION® esiRNA, esiRNA targeting mouse Chn1