EMAIL THIS PAGE TO A FRIEND
Annals of hematology
Plasma clot properties in patients with a mild-to-moderate bleeding tendency of unknown cause.
PMID 25971840
Abstract
In a large proportion of patients with mild bleeding disorders (MBDs) no diagnosis can be established by routine coagulation tests. We investigated whether alterations in plasma clot properties account for MBDs of unknown cause. Ninety-five patients with MBDs of unknown origin and 98 age- and sex-matched healthy controls were investigated. Furthermore, data of 25 patients with a deficiency of factor VIII were analyzed. Plasma clot characteristics in the absence and presence of recombinant tissue plasminogen activator (rtPA) represented by the lag phase, rate of protofibril formation (Vmax), fibrin structure (ΔAbs), time to peak (TTP), half lysis time (t50 and area under the curve (AUC) were measured in turbidometric clot formation and lysis assays. In the fibrinolysis assay, Vmax was lower in patients than in healthy controls. No differences in the other parameters of clot formation and lysis were detected between the groups. There was no clear association of plasma clot properties with the clinical severity of bleeding in patients with MBDs. Patients with known decreased factor VIII levels also showed a lower Vmax. Fibrinogen levels were positively associated with each of the assessed parameters in both groups, with the strongest association with ΔAbs, indicating altered fibrin structure. Factor VIII activity correlated with altered clot characteristics similar to fibrinogen, especially in patients, with the strongest positive correlation to Vmax. This cohort of patients with MBDs of unknown origin showed a lower rate of fibrin formation in the fibrinolysis assay, but otherwise similar plasma clot properties compared to healthy controls.