EMAIL THIS PAGE TO A FRIEND
Metabolomics : Official journal of the Metabolomic Society
GC-MS based targeted metabolic profiling identifies changes in the wheat metabolome following deoxynivalenol treatment.
PMID 25972772