EMAIL THIS PAGE TO A FRIEND
Water research
Electrocoagulation treatment of peat bog drainage water containing humic substances.
PMID 25973580
Abstract
Electrocoagulation (EC) treatment of 100xa0mg/L synthetic wastewater (SWW) containing humic acids was optimized (achieving 90% CODMn and 80% DOC removal efficiencies), after which real peat bog drainage waters (PBDWs) from three northern Finnish peat bogs were also treated. High pollutant removal efficiencies were achieved: Ptot, TS, and color could be removed completely, while Ntot, CODMn, and DOC/TOC removal efficiencies were in the range of 33-41%, 75-90%, and 62-75%, respectively. Al and Fe performed similarly as the anode material. Large scale experiments (1xa0m(3)) using cold (Txa0=xa010-11xa0°C) PBDWs were also conducted successfully, with optimal treatment times of 60-120xa0min (applying current densities of 60-75 A/m(2)). Residual values of Al and Fe (complete removal) were lower than their initial values in the EC-treated PBDWs. Electricity consumption and operational costs in optimum conditions were found to be low and similar for all the waters studied: 0.94 kWh/m(3) and 0.15 €/m(3) for SWW and 0.35-0.70 kWh/m(3) and 0.06-0.12 €/m(3) for the PBDWs (large-scale). Thus, e.g. solar cells could be considered as a power source for this EC application. In conclusion, EC treatment of PBDW containing humic substances was shown to be feasible.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H16752 Humic acid sodium salt, technical grade