EMAIL THIS PAGE TO A FRIEND
ACS applied materials & interfaces
Ratiometric Monitoring of Intracellular Drug Release by an Upconversion Drug Delivery Nanosystem.
PMID 25975535
Abstract
Nanoscale drug delivery systems have been widely investigated due to their well-recognized advantages including controlled delivery of chemotherapeutic agents, enhanced therapeutic effectiveness, and reduced adverse effects compared to conventional chemotherapy with small molecules. However, further progress in the use of nanoscale delivery systems in clinical applications has been hampered by pharmacokinetic studies in biological samples which were associated with significant experimental challenges. Here, we report a rational ratiometric approach to monitor drug release kinetics by quantitatively investigating luminescence resonance energy transfer (LRET) from upconversion nanoparticles to the antitumor drug doxorubicin (DOX). Specifically, DOX molecules within the shell of mesoporous silica-coated upconversion nanoparticles selectively quenched the green emission of upconversion nanoparticles, while the intensity of red emission was essentially unaltered. Consequently, when DOX was gradually released, a steady recovery of green emission was observed. The ability to monitor the intensity ratio of green-to-red luminescence enabled a rational design for real-time investigation of drug delivery release kinetics. Importantly, the internal standard effect of red emission made this ratiometric approach suitable for complex biological microenvironments.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
338869
Ammonium fluoride, ≥99.99% trace metals basis
H4FN