EMAIL THIS PAGE TO A FRIEND
Food chemistry
Extraction and characterisation of pomace pectin from gold kiwifruit (Actinidia chinensis).
PMID 25977029
Abstract
Gold kiwifruit pomace extracted using citric acid, water and enzyme (Celluclast 1.5L) were studied in terms of pectin yield, protein, ash, non-starch polysaccharide, galacturonic acid (GalA), neutral sugar composition, molar mass (Mw), viscosity and degree of branching. Water-extracted pectin was considered closest to its native form. Enzyme extracted pectin showed the highest yield (∼ 4.5%w/w) as compared with the acid and water extraction methods (∼ 3.6-3.8%w/w). Pectin obtained from different extraction methods showed different degree of branching. The Mw and root mean square (RMS) radius varied with the extraction methods with values of 8.4 × 10(5) g/mol and 92 nm, 8.5 × 10(5)g/mol and 102 nm, 6.7 × 10(5) g/mol and 52 nm for acid, water and enzymatic extraction methods, respectively. Similar trend was observed for pectin viscosity, with water-extracted pectin giving a slightly higher viscosity followed by acid and enzyme-extracted pectin. This study showed that gold kiwifruit pomace pectin has potential application in food products.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P9135 Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)