EMAIL THIS PAGE TO A FRIEND
Food chemistry
Effect of high pressure--low temperature treatments on structural characteristics of whey proteins and micellar caseins.
PMID 25977037
Abstract
In this study, structural changes in micellar caseins and whey proteins due to high pressure--low temperature treatments (HPLT) were investigated and compared to changes caused by high pressure treatments at room temperature. Whey protein isolate (WPI) solutions as well as micellar casein (MC) dispersions and mixtures were treated at 500 MPa (pH 7.0 and 5.8) at room temperature, -15 °C and -35 °C. Surface hydrophobicity and accessible thiol groups remained nearly unchanged after HPLT treatments whereas HP treatments at room temperature caused an unfolding of the WPI, resulting in an increase in surface hydrophobicity and exposure of the thiol groups. For HPLT treatments, distinct changes in the secondary structure (increase in the amount of β-sheets) were observed while the tertiary structure remained unchanged. Large flocs, stabilized by hydrophobic interactions and hydrogen bonds, were formed in casein containing samples due to HPLT treatments. Depending on the pH and the applied HPLT treatment parameters, these interactions differed significantly from the interactions determined in native micelles.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A1028
8-Anilino-1-naphthalenesulfonic acid
C16H13NO3S