EMAIL THIS PAGE TO A FRIEND
Genome biology and evolution
Analysis of Five Gene Sets in Chimpanzees Suggests Decoupling between the Action of Selection on Protein-Coding and on Noncoding Elements.
PMID 25977458
Abstract
We set out to investigate potential differences and similarities between the selective forces acting upon the coding and noncoding regions of five different sets of genes defined according to functional and evolutionary criteria: 1) two reference gene sets presenting accelerated and slow rates of protein evolution (the Complement and Actin pathways); 2) a set of genes with evidence of accelerated evolution in at least one of their introns; and 3) two gene sets related to neurological function (Parkinson's and Alzheimer's diseases). To that effect, we combine human-chimpanzee divergence patterns with polymorphism data obtained from target resequencing 20 central chimpanzees, our closest relatives with largest long-term effective population size. By using the distribution of fitness effect-alpha extension of the McDonald-Kreitman test, we reproduce inferences of rates of evolution previously based only on divergence data on both coding and intronic sequences and also obtain inferences for other classes of genomic elements (untranslated regions, promoters, and conserved noncoding sequences). Our results suggest that 1) the distribution of fitness effect-alpha method successfully helps distinguishing different scenarios of accelerated divergence (adaptation or relaxed selective constraints) and 2) the adaptive history of coding and noncoding sequences within the gene sets analyzed is decoupled.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14509
Tin, ≥99%, powder
Sn
265640
Tin, powder, <150 μm, 99.5% trace metals basis
Sn
243434
Tin, granular, 0.425-2.0 mm particle size, ≥99.5%, ACS reagent
Sn
576883
Tin, nanopowder, <150 nm particle size (SEM), ≥99% trace metals basis
Sn
520373
Tin, powder, 10 μm, 99% trace metals basis
Sn
204692
Tin, shot, 99.999% trace metals basis
Sn
217697
Tin, 99.8%, shot, 3 mm
Sn
265632
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sn
265756
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sn
356948
Tin, foil, thickness 0.127 mm, 99.9%
Sn
755702
Tin, powder, -100 mesh, 99.99% trace metals basis
Sn
356956
Tin, wire, diam. 0.5 mm, 99.999% trace metals basis
Sn
GF96022852
Tin, foil, 2m coil, thickness 0.125mm, as rolled, 98.8%
Sn
GF53258182
Tin, foil, 25x25mm, thickness 1.0mm, as rolled, 99.99+%
Sn
GF48212669
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 98.8%
Sn
GF05063228
Tin, foil, 100x100mm, thickness 0.125mm, as rolled, 99.99+%
Sn
GF31638089
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 98.8%
Sn
GF21766068
Tin, foil, 100x100mm, thickness 0.15mm, as rolled, 99.99+%
Sn
GF04538617
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 98.8%
Sn
GF54272336
Tin, foil, 100x100mm, thickness 0.1mm, as rolled, 99.95%
Sn
GF79890889
Tin, foil, 100x100mm, thickness 0.20mm, as rolled, 98.8%
Sn
GF25742943
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 98.8%
Sn
GF11564535
Tin, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Sn
GF45828827
Tin, foil, 100x100mm, thickness 0.5mm, as rolled, 98.8%
Sn
GF93825771
Tin, foil, 100x100mm, thickness 0.5mm, as rolled, 99.99+%
Sn
GF10675470
Tin, foil, 100x100mm, thickness 0.65mm, as rolled, 99.99+%
Sn
GF08231701
Tin, foil, 100x100mm, thickness 0.75mm, as rolled, 99.99+%
Sn
GF39022815
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 98.8%
Sn
GF59159223
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99+%
Sn
GF98532041
Tin, foil, 100x100mm, thickness 1.0mm, as rolled, 99.999%
Sn