EMAIL THIS PAGE TO A FRIEND
Methods (San Diego, Calif.)
Nanometric molecular separation measurements by single molecule photobleaching.
PMID 25980369
Abstract
Although considerable progress has been made in imaging distances in cells below the diffraction limit using FRET and super-resolution microscopy, methods for determining the separation of macromolecules in the 10-50 nm range have been elusive. We have developed fluorophore localisation imaging with photobleaching (FLImP), based on the quantised bleaching of individual protein-bound dye molecules, to quantitate the molecular separations in oligomers and nanoscale clusters. We demonstrate the benefits of using our method in studying the nanometric organisation of the epidermal growth factor receptor in cells.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9891
Doxycycline hyclate, ≥98% (HPLC)
C22H24N2O8 · HCl · 0.5H2O · 0.5C2H6O