EMAIL THIS PAGE TO A FRIEND
Journal of dairy science
Increasing stringiness of low-fat mozzarella string cheese using polysaccharides.
PMID 25981060
Abstract
When fat content of pasta filata cheese is lowered, a loss of fibrous texture occurs and low-fat (LF) mozzarella cheese loses stringiness, making it unsuitable for the manufacture of string cheese. We investigated the use of various polysaccharides that could act as fat mimetics during the stretching and extruding process to aid in protein strand formation and increase stringiness. Low-fat mozzarella cheese curd was made, salted, and then 3.6-kg batches were heated in hot (80°) 5% brine, stretched, and formed into a homogeneous mass. Hot (80°C) slurries of various polysaccharides were then mixed with the hot cheese and formed into LF string cheese using a small piston-driven extruder. Polysaccharides used included waxy corn starch, waxy rice starch, instant tapioca starch, polydextrose, xanthan gum, and guar gum. Adding starch slurries increased cheese moisture content by up to 1.6% but was not effective at increasing stringiness. Xanthan gum functioned best as a fat mimetic and produced LF string cheese that most closely visually resembled commercial string cheese made using low-moisture part skim (LMPS) mozzarella cheese without any increase in moisture content. Extent of stringiness was determined by pulling apart the cheese longitudinally and observing size, length, and appearance of individual cheese strings. Hardness was determined using a modified Warner-Bratzler shear test. When LF string cheese was made using a 10% xanthan gum slurry added at ~1%, increased consumer flavor liking was observed, with scores after 2wk of storage of 6.44 and 6.24 compared with 5.89 for the LF control cheese; although this was lower than an LMPS string cheese that scored 7.27. The 2-wk-old LF string cheeses containing xanthan gum were considered still slightly too firm using a just-about-right (JAR) test, whereas the LMPS string cheese was considered as JAR for texture. With further storage up to 8wk, all of the LF string cheeses softened (JAR score was closer to 3.0); however, much of the stringiness of the LF string cheeses was also lost during storage. We have demonstrated the potential feasibility of increasing stringiness in LF string cheese using polysaccharides with xanthan gum, although further research is needed to develop quantitative methodology for measuring stringiness and to maintain stringiness through the extended refrigerated shelf life needed for string cheese.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
677469
Titanium(IV) oxide, contains 1% Mn as dopant, nanopowder, <100 nm particle size (BET), ≥97%
O2Ti
718467
Titanium(IV) oxide, nanopowder, 21 nm primary particle size (TEM), ≥99.5% trace metals basis
O2Ti
774510
Titanium(IV) oxide, nanowires, diam. × L ~100 nm × 10 μm
O2Ti
774529
Titanium(IV) oxide, nanowires, diam. × L ~10 nm × 10 μm
O2Ti
637254
Titanium(IV) oxide, anatase, nanopowder, <25 nm particle size, 99.7% trace metals basis
O2Ti
232033
Titanium(IV) oxide, anatase, powder, 99.8% trace metals basis
O2Ti
248576
Titanium(IV) oxide, anatase, powder, −325 mesh, ≥99% trace metals basis
O2Ti
634662
Titanium(IV) oxide, mixture of rutile and anatase, nanopowder, <100 nm particle size (BET), 99.5% trace metals basis
O2Ti
700355
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticle, <250 nm particle size (DLS), paste, 53-57 wt. % in diethylene glycol monobutyl ether/ethylene glycol, 99.9% trace metals basis
O2Ti
700347
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <150 nm particle size (volume distribution, DLS), dispersion, 40 wt. % in H2O, 99.5% trace metals basis
O2Ti
700339
Titanium(IV) oxide, mixture of rutile and anatase, nanoparticles, <100 nm particle size, dispersion, 48-52 wt. % in xylene, 99.9% trace metals basis
O2Ti
204730
Titanium(IV) oxide, rutile, 99.995% trace metals basis
O2Ti
204757
Titanium(IV) oxide, rutile, ≥99.98% trace metals basis
O2Ti
635057
Titanium(IV) oxide, rutile, <001>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635049
Titanium(IV) oxide, rutile, <100>, (single crystal substrate), ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
635065
Titanium(IV) oxide, rutile, <110>, single crystal substrate, ≥99.9% trace metals basis, L × W × thickness 10 mm × 10 mm × 0.5 mm
O2Ti
637262
Titanium(IV) oxide, rutile, nanopowder, <100 nm particle size, 99.5% trace metals basis
O2Ti
224227
Titanium(IV) oxide, rutile, powder, <5 μm, ≥99.9% trace metals basis
O2Ti