EMAIL THIS PAGE TO A FRIEND
Toxicology in vitro : an international journal published in association with BIBRA
Impact of graphene oxide on viability of Chinese hamster ovary and mouse hepatoma MH-22A cells.
PMID 25982263
Abstract
The evaluation of the cyto- and bio-compatibility is a critical step in the development of graphene oxide (GO) as a new promising material for in vivo biomedical applications. In this study, we report the impact of GO, with and without the addition of bovine serum albumin, on healthy (Chinese hamster ovary) and a cancer (mouse hepatoma MH-22A) cells viability and the estimation of the intracellular distribution of GO inside the cells in vitro. The viability tests were performed using a colony formation assay. The intracellular distribution of GO was estimated using Raman spectroscopy and imaging. The viability of both cell lines decreased with increasing concentration of graphene oxide (12.5-50.0 μg/ml): in the case of Chinese hamster ovary cells viability decreased from 44% to 11%, in the case of mouse hepatoma MH-22A cells--from 22% to 3%. These cell lines significantly differed in their response to GO and GO-BSA formulations. The results of viability tests correlate with results of atomic force microscopy and Raman spectroscopy and imaging findings. The GO influence on cell morphology changes, cell structure, cells colony growth dynamics and GO accumulation inside the cells was higher in the case of mouse hepatoma MH-22A cells.