EMAIL THIS PAGE TO A FRIEND
Bioelectrochemistry (Amsterdam, Netherlands)
Electrochemical evaluation of poly(3,4-ethylenedioxythiophene) films doped with bacteria based on viability analysis.
PMID 25984659
Abstract
To immobilize viable bacteria on an electrode, we present a novel and straightforward technique that relies on the negative ζ-potentials of bacteria for insertion into conducting polymers as dopants. In the present study, we conducted an electrochemical polymerization of poly(3,4-ethylenedioxythiophene) (PEDOT) doped with various gram-negative bacteria, including Pseudomonas aeruginosa, Escherichia coli, and Shewanella oneidensis. The PEDOT film doped with bacteria indicated a typical redox response, high conductivity, and electrochemical stability. Fluorescence microscopy confirmed that approximately 90% of the bacteria incorporated into the PEDOT film at >0.5 μm in thickness were viable.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
483028
3,4-Ethylenedioxythiophene, 97%
C6H6O2S