EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Successful acquisition of a neutralizing monoclonal antibody against a novel neutrophil-activating peptide, mitocryptide-1.
PMID 25986736
Abstract
Mitocryptide-1 (MCT-1) is a novel neutrophil-activating peptide derived from mitochondrial cytochrome c oxidase subunit VIII, and its physiological role and involvement in various diseases have not yet been elucidated. Generating neutralizing antibodies against the function of MCT-1 is of particular importance for investigating its physiological and pathophysiological roles, because MCT-1 is a fragmented peptide of its mother protein and hence it is very difficult to manipulate its expression level genetically without affecting expression of the mother protein. Here, we report the successful generation of a neutralizing monoclonal antibody (MAb) against MCT-1. This MAb, designated NM1B1, which specifically bound to the region of positions 9-22 of MCT-1, showed concentration-dependent inhibition of MCT-1-induced migration and β-hexosaminidase release in neutrophilic/granulocytic differentiated HL-60 cells. Thus, NM1B1, as a neutralizing MAb against MCT-1, could elucidate not just the physiological regulatory mechanisms of MCT-1 but also its pathophysiological involvement in various inflammatory diseases in vivo.