EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences
Metabolic profile of 2-(2-hydroxypropanamido) benzoic acid in rats by ultra high performance liquid chromatography combined with Fourier transform ion cyclotron resonance mass spectrometry.
PMID 25988429
Abstract
An ultra high performance liquid chromatography combined with Fourier transform ion cyclotron resonance mass spectrometry (UHPLC-FT-ICR-MS) method was developed to investigate the in vivo metabolism of 2-(2-hydroxypropanamido) benzoic acid (HPABA), a marine-derived anti-inflammatory drug, for the first time. Plasma, urine, feces and bile samples were collected from male and female rats after a single intragastric administration of HPABA at a dose of 100mg/kg. Besides the parent drug, a total of 13 metabolites (3 phase I and 10 phase II metabolites) were detected and tentatively identified through comparing their mass spectrometry profiles with those of HPABA. Results demonstrated that metabolic pathways of HPABA in rats included decarboxylation, hydroxylation, dehydrogenation, glucuronidation, glycine conjugation and N-acetyl conjugation. In summary, this work provided valuable information regarding the metabolism of HPABA in rats, which would contribute to better understanding of its safety and mechanism of action.