EMAIL THIS PAGE TO A FRIEND
Nutrients
Hypolipidemic effects and safety of Lactobacillus reuteri 263 in a hamster model of hyperlipidemia.
PMID 25988768
Abstract
We aimed to verify the beneficial effects of probiotic strain Lactobacillus reuteri 263 (Lr263) on hypolipidemic action in hamsters with hyperlipidemia induced by a 0.2% cholesterol and 10% lard diet (i.e., high-cholesterol diet (HCD)). Male Golden Syrian hamsters were randomly divided into two groups: normal (n = 8), standard diet (control), and experimental (n = 32), a HCD. After a two-week induction followed by a six-week supplementation with Lr263, the 32 hyperlipidemic hamsters were divided into four groups (n = 8 per group) to receive vehicle or Lr263 by oral gavage at 2.1, 4.2, or 10.5 × 10(9) cells/kg/day for 6 weeks, designated the HCD, 1X, 2X and 5X groups, respectively. The efficacy and safety of Lr263 supplementation were evaluated by lipid profiles of serum, liver and feces and by clinical biochemistry and histopathology. HCD significantly increased serum levels of total cholesterol (TC), triacylglycerol (TG) cholesterol, high-density lipoprotein cholesterol (HDL-C), and low-density lipoprotein cholesterol (LDL-C), LDL-C/HDL-C ratio, hepatic and fetal TC and TG levels, and degree of fatty liver as compared with controls. Lr263 supplementation dose dependently increased serum HDL-C level and decreased serum TC, TG, LDL-C levels, LDL-C/HDL-C ratio, hepatic TC and TG levels, and fecal TG level. In addition, Lr263 supplementation had few subchronic toxic effects. Lr263 could be a potential agent with a hypolipidemic pharmacological effect.