PloS one
A new therapeutic assessment score for advanced hepatocellular carcinoma patients receiving hepatic arterial infusion chemotherapy.
PMID 25992784
Abstract
Hepatic arterial infusion chemotherapy (HAIC) is an option for treating advanced hepatocellular carcinoma (HCC). Because of the poor prognosis in HAIC non-responders, it is important to identify patients who may benefit from continuous HAIC treatment; however, there are currently no therapeutic assessment scores for this identification. Therefore, we aimed to establish a new therapeutic assessment score for such patients. We retrospectively analyzed 90 advanced HCC patients with elevated baseline alpha-fetoprotein (AFP) and/or des-gamma-carboxy prothrombin (DCP) levels and analyzed various parameters for their possible use as predictors of response and survival. AFP and DCP responses were assessed after half a course of HAIC (2 weeks); a positive-response was defined as a reduction of ≥ 20% from baseline. Multivariate analysis identified DCP response (odds ratio 16.03, p < 0.001) as an independent predictor of treatment response. In multivariate analysis, Child-Pugh class A (hazard ratio [HR] 1.99, p = 0.018), AFP response (HR 2.17, p = 0.007), and DCP response (HR 1.90, p = 0.030) were independent prognostic predictors. We developed an Assessment for Continuous Treatment with HAIC (ACTH) score, including the above 3 factors, which ranged from 0 to 3. Patients stratified into two groups according to this score showed significantly different prognoses (≤ 1 vs. ≥ 2 points: median survival time, 15.1 vs. 8.7 months; p = 0.003). The ACTH score may be useful in the therapeutic assessment of HCC patients receiving HAIC.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O3639
Ondansetron hydrochloride dihydrate, ≥98% (HPLC), powder
C18H19N3O · HCl · 2H2O