EMAIL THIS PAGE TO A FRIEND
FEMS microbiology letters
Identification of N1-acetylnorspermidine in Vibrio parahaemolyticus and an enzyme activity responsible for its formation.
PMID 2599358
Abstract
N1-Acetylnorspermidine [CH3CONH(CH2)3 NH(CH2)3NH3] was identified in Vibrio parahaemolyticus, which contains norspermidine as a major polyamine. This is the first example for the natural occurrence of monoacetylated unusual polyamine. The N1-acetylnorspermidine content was the highest 4 h after inoculation. Incubation of norspermidine and acetyl CoA with a cell extract from V. parahaemolyticus produced N1-acetylnorspermidine. A remarkable increase in specific activity of the acetyltransferase was observed at the exponential phase of growth. Spermidine also served as a substrate for the enzyme, with the formation of two isomers of the acetylspermidines (N1-acetylspermidine was predominant), but the reaction rate was less than 50% of that with norspermidine. These results suggest that norspermidine in V. parahaemolyticus may be associated with the cell growth and its role may be controlled through acetylation, as reported for spermidine in Escherichia coli.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
14520
Bis(3-aminopropyl)amine, purum, ≥97.0% (GC)
C6H17N3
I1006
Bis(3-aminopropyl)amine, 98%
C6H17N3