EMAIL THIS PAGE TO A FRIEND
American journal of physiology. Regulatory, integrative and comparative physiology
Arterial baroreceptor reflex control of renal sympathetic nerve activity following chronic myocardial infarction in male, female, and ovariectomized female rats.
PMID 25994953
Abstract
There is controversy regarding whether the arterial baroreflex control of renal sympathetic nerve activity (SNA) in heart failure is altered. We investigated the impact of sex and ovarian hormones on changes in the arterial baroreflex control of renal SNA following a chronic myocardial infarction (MI). Renal SNA and arterial pressure were recorded in chloralose-urethane anesthetized male, female, and ovariectomized female (OVX) Wistar rats 6-7 wk postsham or MI surgery. Animals were grouped according to MI size (sham, small and large MI). Ovary-intact females had a lower mortality rate post-MI (24%) compared with both males (38%) and OVX (50%) (P < 0.05). Males and OVX with large MI, but not small MI, displayed an impaired ability of the arterial baroreflex to inhibit renal SNA. As a result, the male large MI group (49 ± 6 vs. 84 ± 5% in male sham group) and OVX large MI group (37 ± 3 vs. 75 ± 5% in OVX sham group) displayed significantly reduced arterial baroreflex range of control of normalized renal SNA (P < 0.05). In ovary-intact females, arterial baroreflex control of normalized renal SNA was unchanged regardless of MI size. In males and OVX there was a significant, positive correlation between left ventricle (LV) ejection fraction and arterial baroreflex range of control of normalized renal SNA, but not absolute renal SNA, that was not evident in ovary-intact females. The current findings demonstrate that the arterial baroreflex control of renal SNA post-MI is preserved in ovary-intact females, and the state of left ventricular dysfunction significantly impacts on the changes in the arterial baroreflex post-MI.